Anti-ICAM3 Rabbit Recombinant Antibody [clone: ICAM3/2873R]
ANTIA248913-100
New Product
- Antikörper-Typ:Primär
- Antigen-Name:CD50
- Antigen-Symbol:ICAM3
- Klonalität:Recombinant
- Klon:ICAM3/2873R
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human
- Form:Liquid
- Gen-ID:UniprotID# P32942
- Antigen-Synonyme:ICAM 3|ICAM-3|CDW 50|ICAM3|Intercellular adhesion molecule 3|CDw50|ICAM3_HUMAN|ICAM-R|ICAMR
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:110 - 160 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:200 µg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ICAM3 protein
- Validierte Anwendungen:IHC-P
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Recombinant rabbit monoclonal [ICAM3/2873R] antibody to ICAM3 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ICAM3
Clonality: Recombinant
Clone: ICAM3/2873R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human