Anti-G-CSF Mouse Recombinant Antibody [clone: rCSF3/900]
Artikelnr. ANTIA248277-100
Lieferant: ANTIBODIES.COM
New Product
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:C17orf33
- Antigen-Symbol:G-CSF
- Klonalität:Recombinant
- Klon:rCSF3/900
- Konjugation:Unconjugated
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG1 kappa
- Reaktivität:Macaque,Human
- Form:Liquid
- Gen-ID:UniprotID# P09919
- Antigen-Synonyme:Colony stimulating factor 3 (granulocyte)|CSF3_HUMAN|Colony stimulating factor 3|Filgrastim|CSF3|CSF3OS|Csfg|CSF beta|CSF 3
- Lagerungspuffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:19 kDa
- Lagertemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Konzentration:200 µg/ml
- Transporttemperatur:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CSF3 protein
- Validierte Anwendungen:IHC-P
- Reinigung:Protein A/G chromatography
- Packung:Plastic vial
- VE:100 µG
Spezifikationen
Über diesen Artikel
Recombinant mouse monoclonal [rCSF3/900] antibody to G-CSF for IHC-P with samples derived from human and macaque monkey.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: G-CSF
Clonality: Recombinant
Clone: rCSF3/900
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Macaque Monkey