Order Entry
Austria
ContactUsLinkComponent
Anti-CCR8 Rabbit Recombinant Antibody [clone: DM197]
Anti-CCR8 Rabbit Recombinant Antibody [clone: DM197]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-CCR8 Rabbit Recombinant Antibody [clone: DM197]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA318512-500
  • ANTIA318512-100
  • Antigen-Symbol:
  • CCR8
  • CCR8
  • Antigen-Name:
  • C C chemokine receptor type 8
  • C C chemokine receptor type 8
  • Antigen-Synonyme:
  • C-C CKR-8|CC chemokine receptor CHEMR1|C C CKR 8|CC CKR 8|CC chemokine receptor 8|C-C chemokine receptor type 8|C C motif chemokine receptor 8|CC chemokine receptor type 8|CC-CKR-8
  • C-C CKR-8|CC chemokine receptor CHEMR1|C C CKR 8|CC CKR 8|CC chemokine receptor 8|C-C chemokine receptor type 8|C C motif chemokine receptor 8|CC chemokine receptor type 8|CC-CKR-8
  • Antikörper-Typ:
  • Primär
  • Primär
  • Klonalität:
  • Recombinant
  • Recombinant
  • Konjugation:
  • Unconjugated
  • Unconjugated
  • Klon:
  • DM197
  • DM197
  • Reaktivität:
  • Human
  • Human
  • Wirt:
  • Rabbit
  • Rabbit
  • Gen-ID:
  • UniprotID# P51685
  • UniprotID# P51685
  • Isotyp:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Durchflusszytometrie:
  • Yes
  • Yes
  • Form:
  • lyophilised
  • lyophilised
  • Reinigung:
  • Affinity chromatography
  • Affinity chromatography
  • Konzentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Lagerungspuffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lagertemperatur:
  • Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Transporttemperatur:
  • Shipped on blue ice at +4 °C
  • Shipped on blue ice at +4 °C

Specifications

Frequently Bought Together