Anti-Eph Receptor A3 Rabbit Recombinant Antibody [clone: DM162]
Lieferant: ANTIBODIES.COM
New Product
Zertifikate
Über diesen Artikel
Recombinant rabbit monoclonal [DM162] antibody to Eph Receptor A3 for ELISA and Flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: Eph Receptor A3
Clonality: Recombinant
Clone: DM162
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spezifikationen
- Catalog No:
- ANTIA318546-500
- ANTIA318546-100
- Antigen-Symbol:
- Eph Receptor A3
- Eph Receptor A3
- Antigen-Name:
- AW492086
- AW492086
- Antigen-Synonyme:
- EC 2.7.10.1|EPHA3|Ephrin receptor EphA3|EPHA3_HUMAN|EPH-like tyrosine kinase 1|Eph receptor A3|Cek4|End3|EPH-like kinase 4
- EC 2.7.10.1|EPHA3|Ephrin receptor EphA3|EPHA3_HUMAN|EPH-like tyrosine kinase 1|Eph receptor A3|Cek4|End3|EPH-like kinase 4
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Recombinant
- Recombinant
- Konjugation:
- Unconjugated
- Unconjugated
- Klon:
- DM162
- DM162
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- UniprotID# P29320
- UniprotID# P29320
- Isotyp:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Form:
- lyophilised
- lyophilised
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Konzentration:
- Reconstitution dependent
- Reconstitution dependent
- Lagerungspuffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lagertemperatur:
- Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Transporttemperatur:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Spezifikationen
Frequently Bought Together