Spezifikationen
- VE:96 Tests
- Testdauer:Multiple steps
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:RED
- Bezeichnung:Human RED ELISA kit
- Probentyp:Serum, plasma, tissue homogenates and other biological fluids
- Kreuzreaktivität:Human RED ELISA Kit exhibits high specificity and excellent specificity for the detection of human RED. No significant cross-reactivity or interference between RED and analogues was observed.
- Erkennungsmethode:Colorimetric
- Time to Results:4 h 30 min
- Testprinzip:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:31,25 - 2000 pg/ml
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Empfindlichkeit:18,75 pg/ml
- Regulatory status:RUO
Spezifikationen
Über diesen Artikel
Human RED ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RED in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA Kit
- Detection range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human RED ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human RED in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for RED has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RED present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-RED Antibody, which binds the captured RED present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of RED captured in each well. The concentration of RED can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.