Human TRPV2 ELISA Kit
Product Details & Documents
Human TRPV2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TRPV2 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 100 to 1600 ng/L
- Sensitivity: 7,98 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human TRPV2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TRPV2 in An antibody specific for TRPV2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TRPV2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-TRPV2 antibody, which also binds the TRPV2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-TRPV2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-TRPV2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of TRPV2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of TRPV2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- VE:96 Tests
- Bezeichnung:Human TRPV2 ELISA kit
- Empfindlichkeit:7,98 ng/L
- Reaktivität der primären Antikörper:Human
- Shelf life:Store for 6 months at 4 °C
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ANTIA312902-96
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Format:Pre-coated
- Target protein:TRPV2
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, plasma or other biological fluids
- Erfassungsbereich:100 - 1600 ng/L
- Regulatory status:RUO
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



