Specifications
- VE:96 Tests
- Testdauer:One step
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:Aryl Hydrocarbon Receptor
- Bezeichnung:Human Aryl Hydrocarbon receptor ELISA kit
- Probentyp:Serum, plasma or other biological fluids
- Erkennungsmethode:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:75 - 1200 ng/L
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Empfindlichkeit:5,25 ng/L
- Regulatory status:RUO
Specifications
About this item
Human Aryl Hydrocarbon receptor ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Aryl Hydrocarbon receptor in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Aryl Hydrocarbon receptor ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Aryl Hydrocarbon receptor in serum, plasma or other biological fluids. An antibody specific for Aryl Hydrocarbon receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Aryl Hydrocarbon receptor present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Aryl Hydrocarbon receptor antibody, which also binds the Aryl Hydrocarbon receptor present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Aryl Hydrocarbon receptor antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Aryl Hydrocarbon receptor antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Aryl Hydrocarbon receptor captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Aryl Hydrocarbon receptor in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.