Order Entry
Austria
ContactUsLinkComponent
 
Mouse S100 beta ELISA Kit
  ANTIA312798-96
New Product
 :  
undefined
Mouse S100 beta ELISA Kit
  ANTIA312798-96
New Product
 :  

 

  • VE:
    96 Tests
  • Testdauer:
    One step
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Mouse
  • Target protein:
    S100 beta
  • Bezeichnung:
    Mouse S100 beta ELISA kit
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    7,5 - 120 pg/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Empfindlichkeit:
    0,23 pg/ml
  • Regulatory status:
    RUO

 

 

Mouse S100 beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse S100 beta in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 7,5 to 120 pg/ml
  • Sensitivity: 0,23 pg/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse S100 beta ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse S100 beta in serum, plasma or other biological fluids. An antibody specific for S100 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the S100 beta present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-S100 beta antibody, which also binds the S100 beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-S100 beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-S100 beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of S100 beta captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of S100 beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.