Order Entry
Austria
ContactUsLinkComponent
Human Apolipoprotein CII/ApoC-II ELISA Kit
Human Apolipoprotein CII/ApoC-II ELISA Kit
  ANTIA312727-96
New Product
 :  
undefined
Human Apolipoprotein CII/ApoC-II ELISA Kit
  ANTIA312727-96
New Product
 :  

 

  • VE:
    96 Tests
  • Testdauer:
    One step
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    Apolipoprotein CII/ApoC-II
  • Bezeichnung:
    Human Apolipoprotein CII/ApoC-II ELISA kit
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    125 - 2000 ng/L
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Empfindlichkeit:
    9,68 ng/L
  • Regulatory status:
    RUO

 

 

Human Apolipoprotein CII/ApoC-II ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Apolipoprotein CII/ApoC-II in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Apolipoprotein CII/ApoC-II ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Apolipoprotein CII/ApoC-II in serum, plasma or other biological fluids. An antibody specific for Apolipoprotein CII/ApoC-II has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Apolipoprotein CII/ApoC-II present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Apolipoprotein CII/ApoC-II antibody, which also binds the Apolipoprotein CII/ApoC-II present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Apolipoprotein CII/ApoC-II antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Apolipoprotein CII/ApoC-II antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Apolipoprotein CII/ApoC-II captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Apolipoprotein CII/ApoC-II in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.