Order Entry
Austria
ContactUsLinkComponent
 
Human SLC24A6 ELISA Kit
Catalog # ANTIA302746-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human SLC24A6 ELISA Kit
Catalog # ANTIA302746-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    SLC24A6
  • Bezeichnung:
    Human SLC24A6 ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Kreuzreaktivität:
    Human SLC24A6 ELISA Kit exhibits high specificity and excellent specificity for the detection of human SLC24A6. No significant cross-reactivity or interference between SLC24A6 and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    15,625 - 1000 pg/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    9,375 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human SLC24A6 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SLC24A6 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human SLC24A6 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SLC24A6 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SLC24A6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SLC24A6 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SLC24A6 Antibody, which binds the captured SLC24A6 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SLC24A6 captured in each well. The concentration of SLC24A6 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.