- VE:96 Tests
- Testdauer:Multiple steps
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:OSCP1
- Bezeichnung:Human OSCP1 ELISA kit
- Probentyp:Serum, plasma, tissue homogenates, and other biological fluids
- Kreuzreaktivität:Human OSCP1 ELISA Kit exhibits high specificity and excellent specificity for the detection of human OSCP1. No significant cross-reactivity or interference between OSCP1 and analogues was observed.
- Erkennungsmethode:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:31,25 - 2000 pg/ml
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Empfindlichkeit:18,75 pg/ml
- Regulatory status:RUO
Human OSCP1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human OSCP1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human OSCP1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human OSCP1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for OSCP1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the OSCP1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-OSCP1 Antibody, which binds the captured OSCP1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of OSCP1 captured in each well. The concentration of OSCP1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.