Specifications
- VE:96 Tests
- Testdauer:One step
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:Calpain Small Subunit 1
- Bezeichnung:Human Calpain small subunit 1 ELISA kit
- Probentyp:Serum, plasma or other biological fluids
- Erkennungsmethode:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:0,5 - 8 ng/ml
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Empfindlichkeit:0,02 ng/ml
- Regulatory status:RUO
Specifications
About this item
Human Calpain Small Subunit 1 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Calpain Small Subunit 1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Calpain Small Subunit 1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Calpain Small Subunit 1 in serum, plasma or other biological fluids. An antibody specific for Calpain Small Subunit 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Calpain Small Subunit 1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Calpain Small Subunit 1 antibody, which also binds the Calpain Small Subunit 1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Calpain Small Subunit 1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Calpain Small Subunit 1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Calpain Small Subunit 1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Calpain Small Subunit 1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.