- VE:96 Tests
- Testdauer:One step
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Mouse
- Target protein:Retinoic acid Receptor beta
- Bezeichnung:Mouse Retinoic acid receptor beta ELISA kit
- Probentyp:Serum, plasma or other biological fluids
- Erkennungsmethode:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:1,5 - 24 ng/ml
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Empfindlichkeit:0,042 ng/ml
- Regulatory status:RUO
Mouse Retinoic acid Receptor beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Retinoic acid Receptor beta in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 1,5 to 24 ng/ml
- Sensitivity: 0,042 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Retinoic acid Receptor beta ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Retinoic acid Receptor beta in serum, plasma or other biological fluids. An antibody specific for Retinoic acid Receptor beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Retinoic acid Receptor beta present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Retinoic acid Receptor beta antibody, which also binds the Retinoic acid Receptor beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Retinoic acid Receptor beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Retinoic acid Receptor beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Retinoic acid Receptor beta captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Retinoic acid Receptor beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.