Order Entry
Austria
ContactUsLinkComponent
Mouse Retinoic acid Receptor gamma ELISA Kit
Mouse Retinoic acid Receptor gamma ELISA Kit
Catalog # ANTIA314112-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Mouse Retinoic acid Receptor gamma ELISA Kit
Catalog # ANTIA314112-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • VE:
    96 Tests
  • Testdauer:
    One step
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Mouse
  • Target protein:
    Retinoic acid Receptor gamma
  • Bezeichnung:
    Mouse Retinoic acid receptor gamma ELISA kit
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    1 - 16 ng/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Empfindlichkeit:
    0,026 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Retinoic acid Receptor gamma ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Retinoic acid Receptor gamma in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1 to 16 ng/ml
  • Sensitivity: 0,026 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Retinoic acid Receptor gamma ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Retinoic acid Receptor gamma in serum, plasma or other biological fluids. An antibody specific for Retinoic acid Receptor gamma has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Retinoic acid Receptor gamma present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Retinoic acid Receptor gamma antibody, which also binds the Retinoic acid Receptor gamma present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Retinoic acid Receptor gamma antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Retinoic acid Receptor gamma antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Retinoic acid Receptor gamma captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Retinoic acid Receptor gamma in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.