Order Entry
Austria
ContactUsLinkComponent
Human KLF5 ELISA Kit
Human KLF5 ELISA Kit
Catalog # ANTIA314074-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human KLF5 ELISA Kit
Catalog # ANTIA314074-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • VE:
    96 Tests
  • Testdauer:
    One step
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    KLF5
  • Bezeichnung:
    Human KLF5 ELISA kit
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    150 - 2400 pmol/L
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Empfindlichkeit:
    5,21 pmol/L
  • Regulatory status:
    RUO

Specifications

About this item

Human KLF5 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KLF5 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 to 2400 pmol/L
  • Sensitivity: 5,21 pmol/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human KLF5 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human KLF5 in serum, plasma or other biological fluids. An antibody specific for KLF5 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the KLF5 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-KLF5 antibody, which also binds the KLF5 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-KLF5 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-KLF5 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of KLF5 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of KLF5 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.