Specifications
- VE:96 Tests
- Testdauer:One step
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:ZO1 Tight Junction Protein
- Bezeichnung:Human ZO1 Tight junction protein ELISA kit
- Probentyp:Serum, plasma or other biological fluids
- Erkennungsmethode:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:200 - 3200 ng/L
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Empfindlichkeit:9,46 ng/L
- Regulatory status:RUO
Specifications
About this item
Human ZO1 Tight Junction Protein ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ZO1 Tight Junction Protein in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 200 to 3200 ng/L
- Sensitivity: 9,46 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human ZO1 Tight Junction Protein ELISA kit (A313822) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ZO1 Tight Junction Protein in An antibody specific for ZO1 Tight Junction Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ZO1 Tight Junction Protein present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-ZO1 Tight Junction Protein antibody, which also binds the ZO1 Tight Junction Protein present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-ZO1 Tight Junction Protein antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-ZO1 Tight Junction Protein antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of ZO1 Tight Junction Protein captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of ZO1 Tight Junction Protein in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.