Express Eingabe
Austria
AuftragsverfolgungKontakt
Human RAB27B ELISA Kit | Avantor

Human RAB27B ELISA Kit

Human RAB27B ELISA kit, 96 Tests
Artikelnr. ANTIA313468-96
Anbieter:   Antibodies.com
Human RAB27B ELISA Kit
Artikelnr. ANTIA313468-96
Anbieter:  Antibodies.com
CAS Nummer:  
Human RAB27B ELISA Kit
Human RAB27B ELISA Kit

Human RAB27B ELISA kit, 96 Tests

CAS Nummer:  
Preis
565,00 €
Einloggen für Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Human RAB27B ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RAB27B in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 9,04 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human RAB27B ELISA kit (A313468) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human RAB27B in serum, plasma or other biological fluids. An antibody specific for RAB27B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RAB27B present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-RAB27B antibody, which also binds the RAB27B present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-RAB27B antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-RAB27B antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of RAB27B captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of RAB27B in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Bestellinformation: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Spezifikationen

Spezifikationen

  • VE:
    96 Tests
  • Bezeichnung:
    Human RAB27B ELISA kit
  • Empfindlichkeit:
    9,04 ng/L
  • Reaktivität der primären Antikörper:
    Human
  • Shelf life:
    Store for 6 months at 4 °C
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA313468-96
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Format:
    Pre-coated
  • Target protein:
    RAB27B
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erfassungsbereich:
    125 - 2000 ng/L
  • Regulatory status:
    RUO

Spezifikationen

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.