Order Entry
Austria
ContactUsLinkComponent
 
Human Meprin Alpha ELISA Kit
  ANTIA313106-96
New Product
 :  
undefined
Human Meprin Alpha ELISA Kit
  ANTIA313106-96
New Product
 :  

 

  • VE:
    96 Tests
  • Testdauer:
    One step
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    Meprin alpha
  • Bezeichnung:
    Human Meprin alpha ELISA kit
  • Probentyp:
    Serum, plasma or other biological fluids
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    150 - 2400 ng/L
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    40 μl
  • Empfindlichkeit:
    9,29 ng/L
  • Regulatory status:
    RUO

 

 

Human Meprin alpha ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Meprin alpha in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9,29 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Meprin alpha ELISA kit (A313106) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Meprin alpha in serum, plasma or other biological fluids. An antibody specific for Meprin alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Meprin alpha present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Meprin alpha antibody, which also binds the Meprin alpha present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Meprin alpha antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Meprin alpha antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Meprin alpha captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Meprin alpha in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.