- VE:96 Tests
- Testdauer:One step
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:ARFGAP1
- Bezeichnung:Human ARFGAP1 ELISA kit
- Probentyp:Serum, plasma or other biological fluids
- Erkennungsmethode:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:62,5 - 1000 ng/L
- Lagertemperatur:4 °C
- Probenvolumen:40 μl
- Empfindlichkeit:2,19 ng/L
- Regulatory status:RUO
Human ARFGAP1 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ARFGAP1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human ARFGAP1 ELISA kit (A313767) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ARFGAP1 in serum, plasma or other biological fluids. An antibody specific for ARFGAP1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ARFGAP1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-ARFGAP1 antibody, which also binds the ARFGAP1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-ARFGAP1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-ARFGAP1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of ARFGAP1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of ARFGAP1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.