Order Entry
Austria
ContactUsLinkComponent
Human Ceramide Glucosyltransferase ELISA Kit
Human Ceramide Glucosyltransferase ELISA Kit
Catalog # ANTIA78952-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Ceramide Glucosyltransferase ELISA Kit
Catalog # ANTIA78952-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Human
  • Target protein:
    Ceramide Glucosyltransferase
  • Bezeichnung:
    Human ceramide glucosyltransferase ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kreuzreaktivität:
    Human Ceramide Glucosyltransferase ELISA kit exhibits high specificity and excellent specificity for the detection of human Ceramide Glucosyltransferase. No significant cross-reactivity or interference between Ceramide Glucosyltransferase and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Testprinzip:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    0,781 - 50 ng/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    0,469 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Ceramide Glucosyltransferase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Ceramide Glucosyltransferase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0,781 - 50 ng/ml
  • Sensitivity: 0,469 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Ceramide Glucosyltransferase ELISA kit (A78952) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Ceramide Glucosyltransferase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Ceramide Glucosyltransferase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Ceramide Glucosyltransferase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Ceramide Glucosyltransferase Antibody, which binds the captured Ceramide Glucosyltransferase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Ceramide Glucosyltransferase captured in each well. The concentration of Ceramide Glucosyltransferase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.