Order Entry
Austria
ContactUsLinkComponent
 
Rat Daxx ELISA Kit
Catalog # ANTIA79256-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Rat Daxx ELISA Kit
Catalog # ANTIA79256-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Rat
  • Target protein:
    Daxx
  • Bezeichnung:
    Rat Daxx ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kreuzreaktivität:
    Rat Daxx ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Daxx. No significant cross-reactivity or interference between Daxx and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Testprinzip:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    78,125 - 5000 pg/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    46,875 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat Daxx ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Daxx in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Daxx ELISA Kit (A79256) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Daxx in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Daxx has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Daxx present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Daxx Antibody, which binds the captured Daxx present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Daxx captured in each well. The concentration of Daxx can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.