Order Entry
Austria
ContactUsLinkComponent
 
Rat beta 2 Defensin/BD-2 ELISA Kit
  ANTIA79268-96
New Product
 :  
undefined
Rat beta 2 Defensin/BD-2 ELISA Kit
  ANTIA79268-96
New Product
 :  

 

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Rat
  • Target protein:
    beta 2 Defensin/BD-2
  • Bezeichnung:
    Rat beta 2 defensin/BD-2 ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates and other biological fluids
  • Kreuzreaktivität:
    Rat beta 2 Defensin/BD-2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat beta 2 Defensin/BD-2. No significant cross-reactivity or interference between beta 2 Defensin/BD-2 and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Testprinzip:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    0,313 - 20 ng/ml
  • Lagertemperatur:
    4 °C
  • Probenvolumen:
    100 μl
  • Empfindlichkeit:
    0,188 ng/ml
  • Regulatory status:
    RUO

 

 

Rat beta 2 Defensin/BD-2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat beta 2 Defensin/BD-2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat beta 2 Defensin / BD-2 ELISA Kit (A79268) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat beta 2 Defensin / BD-2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for beta 2 Defensin / BD-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the beta 2 Defensin / BD-2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-beta 2 Defensin / BD-2 Antibody, which binds the captured beta 2 Defensin / BD-2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of beta 2 Defensin / BD-2 captured in each well. The concentration of beta 2 Defensin / BD-2 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.