Specifications
- VE:96 Tests
- Testdauer:Multiple steps
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Mouse
- Target protein:c-Kit
- Bezeichnung:Mouse c-kit ELISA kit
- Probentyp:Serum, plasma, tissue homogenates and other biological fluids
- Kreuzreaktivität:Mouse c-Kit ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse c-Kit. No significant cross-reactivity or interference between c-Kit and analogues was observed.
- Erkennungsmethode:Colorimetric
- Time to Results:4 h 30 min
- Testprinzip:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:0,156 - 10 ng/ml
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Empfindlichkeit:0,094 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse c-kit ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse c-kit in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA kit
- Detection Range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Mouse c-kit ELISA kit (A78768) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse c-kit in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for c-kit has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the c-kit present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-c-kit Antibody, which binds the captured c-kit present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of c-kit captured in each well. The concentration of c-kit can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.