Spezifikationen
- VE:96 Tests
- Testdauer:Multiple steps
- Testtyp:Competitive
- Format:Pre-coated
- Reaktivität der primären Antikörper:Other
- Target protein:Prostacyclin 2
- Bezeichnung:Prostacyclin 2 ELISA kit
- Probentyp:Serum, plasma, tissue homogenates, and other biological fluids
- Kreuzreaktivität:Prostacyclin 2 ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Prostacyclin 2. No significant cross-reactivity or interference between Prostacyclin 2 and analogues was observed.
- Erkennungsmethode:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:31,25 - 2000 pg/ml
- Lagertemperatur:4 °C
- Empfindlichkeit:18,75 pg/ml
- Regulatory status:RUO
Spezifikationen
Über diesen Artikel
Prostacyclin 2 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Prostacyclin 2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Prostacyclin 2 ELISA kit (A303901) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Prostacyclin 2 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Prostacyclin 2 antigen. During the incubation, Prostacyclin 2 present in the samples or standards competes with the fixed amount of immobilised Prostacyclin 2 for binding sites on the Biotinylated Anti-Prostacyclin 2 Antibody. The more Prostacyclin 2 present in a sample or standard, the less Biotinylated Anti-Prostacyclin 2 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Prostacyclin 2 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Prostacyclin 2 present in each sample or standard. The concentration of Prostacyclin 2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.