Order Entry
Austria
ContactUsLinkComponent
 
Mouse UCP1 ELISA Kit
  ANTIA303388-96
New Product
 :  
undefined
Mouse UCP1 ELISA Kit
  ANTIA303388-96
New Product
 :  

 

  • VE:
    96 Tests
  • Testdauer:
    Multiple steps
  • Testtyp:
    Sandwich
  • Format:
    Pre-coated
  • Reaktivität der primären Antikörper:
    Mouse
  • Target protein:
    UCP1
  • Bezeichnung:
    Mouse UCP1 ELISA kit
  • Probentyp:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Kreuzreaktivität:
    Mouse UCP1 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse UCP1. No significant cross-reactivity or interference between UCP1 and analogues was observed.
  • Erkennungsmethode:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Erfassungsbereich:
    78,125 - 5000 pg/ml
  • Lagertemperatur:
    4 °C
  • Empfindlichkeit:
    46,875 pg/ml
  • Regulatory status:
    RUO

 

 

Mouse UCP1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse UCP1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse UCP1 ELISA kit (A303388) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse UCP1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for UCP1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the UCP1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-UCP1 Antibody, which binds the captured UCP1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of UCP1 captured in each well. The concentration of UCP1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.