Specifications
- VE:96 Tests
- Testdauer:Multiple steps
- Testtyp:Sandwich
- Format:Pre-coated
- Reaktivität der primären Antikörper:Human
- Target protein:OCT2
- Bezeichnung:Human OCT2 ELISA kit
- Probentyp:Serum, plasma, tissue homogenates and other biological fluids
- Kreuzreaktivität:Human OCT2 ELISA Kit exhibits high specificity and excellent specificity for the detection of human OCT2. No significant cross-reactivity or interference between OCT2 and analogues was observed.
- Erkennungsmethode:Colorimetric
- Time to Results:4 h 30 min
- Testprinzip:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Erfassungsbereich:31,25 - 2000 pg/ml
- Lagertemperatur:4 °C
- Probenvolumen:100 μl
- Empfindlichkeit:18,75 pg/ml
- Regulatory status:RUO
Specifications
About this item
Human OCT2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human OCT2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human OCT2 ELISA Kit (A75740) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human OCT2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for OCT2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the OCT2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-OCT2 Antibody, which binds the captured OCT2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of OCT2 captured in each well. The concentration of OCT2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.