Amplitube™ Cap Strips for Well Plates
Catalog # SIMPT323-1N
Supplier: Simport Scientific
New Product
Specifications
- Farbe:Natur
- Bezeichnung:Domed 8-strip cap
- VE:125
Specifications
About this item
Amplitube™ domed cap strips ensure a perfect closure and a positive seal during thermal cycling and prevent evaporation.
- 8 domed cap strips
- Made of FDA compliant polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Leakproof/evaporation tested
Packaging: Packaged in tamperproof resealable bags.