ContactUsLinkComponent
Human Recombinant Apolipoprotein H, His Tag
  STMC100-1332
 :  
undefined
Human Recombinant Apolipoprotein H, His Tag
  STMC100-1332
 :  
  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Spezies:
    Human
  • Seq:
    1286
  • Proteinsynonym:
    B2G1|β2GP1|β2GPI|BG
  • Protein/Peptid-Name:
    Apolipoprotein H
  • Purity:
    ≥93%
Apolipoprotein H (apo H) has been shown to promote the coagulation of blood platelets by inhibiting thrombomodulin complex and inactivating protein C (Keeling et al.), but can also act as an anticoagulant by binding thrombin and inhibiting its procoagulant effects (Pozzi et al.). Belonging to the lipid-binding apolipoprotein family, within the lipocalin superfamily, apo H is a protein constituent of plasma, with a high affinity for negatively charged phospholipids. The structure of apo H reveals four N-terminal complement control protein (CCP) modules, also known as 'sushi' domains, and a distinct fifth C-terminal domain with four antiparallel beta sheets, two alpha-helices, and an extended loop (Schwarzenbacher et al.). Apo H is the main antigen implicated in antiphospholipid syndrome (APS), an autoimmune condition involving pregnancy complications and vascular thrombosis (Brusch). Studies have also reported that Apo H is involved in the progression of atherosclerosis (Harats and George). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, apolipoprotein H from STEMCELL comes lyophilised with ≥93% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').