ContactUsLinkComponent
CMV (IE2) Peptide Pool
Artikelnr. STMC100-1412
CAS-Nummer:  
undefined
CMV (IE2) Peptide Pool
Artikelnr. STMC100-1412
CAS-Nummer:  
Spezifikationen
  • VE:
    1 AU
  • Protein/Peptid-Name:
    CMV (IE2) Peptide Pool
  • Purity:
    Average 70%
Spezifikationen
Über diesen Artikel
Stimulating T cells with CMV (IE-2) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. CMV (IE-2) Peptide pool is a lyophilised mixture of 143 peptides from immediate early protein 2 (IE2) of human cytomegalovirus (CMV; strain AD169), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 580 on IE2. IE2 is critical for viral infection and for the expression of CMV early genes (Marchini et al.). It also controls transcription initiation for a subset of early-late and late viral promoters via RNA polymerase II (Li et al.). IE2, along with IE1, may contribute to the survival of CMV-infected cells by blocking tumor necrosis factor alpha (TNF-α)-induced apoptosis (Zhu et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').