ContactUsLinkComponent
SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 Mutation Peptide Pool
Artikelnr. STMC100-1382
CAS-Nummer:  
undefined
SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 Mutation Peptide Pool
Artikelnr. STMC100-1382
CAS-Nummer:  
Spezifikationen
  • VE:
    1 AU
  • Protein/Peptid-Name:
    SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 Mutation Peptide Pool
  • Purity:
    Average 70%
Spezifikationen
Über diesen Artikel
Stimulating T cells with SARS-CoV-2 (Spike Protein) Omicron/B.1.1.529 Mutation Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. A lyophilised mixture of 80 peptides of the spike glycoprotein of SARS-CoV-2 Omicron variant (B.1.1.529), these peptides selectively cover the mutated regions in the spike protein of the SARS-CoV-2 Omicron variant, compared to wild-type. Through interactions with the angiotensin-converting enzyme 2 (ACE2) receptor, the spike protein enables the virus to attach to the cell membrane, and plays a critical role in viral entry (Hoffmann et al.; Walls et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').