AuftragsverfolgungKontakt
STEMdiff™ Hepatocyte Kit | Avantor
STEMdiff™ Hepatocyte Kit
STEMdiff™ Hepatocyte kit, 1 KIT
Artikelnr. STMC100-0520
Marketsource
STEMdiff™ Hepatocyte Kit
Artikelnr. STMC100-0520
Marketsource
CAS Nummer:  
STEMdiff™ Hepatocyte Kit
STEMdiff™ Hepatocyte Kit
STEMdiff™ Hepatocyte kit, 1 KIT
CAS Nummer:  
Preis
1 870,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Serum-free differentiation kit for the generation of hepatocyte-like cells from human PSCs.
Efficiently generate hepatocyte-like cells (HLCs) that express several key liver-specific markers, including albumin and alpha1-antitrypsin. The STEMdiff™ Hepatocyte Kit provides a reliable source of HLCs for experiments by reproducibly differentiating human pluripotent stem cells (hPSCs) into HLCs across a variety of hPSC lines. The serum-free formulation minimises experimental variability by reducing undefined components and enables robust differentiation of HLC cultures. HLCs generated using STEMdiff™ Hepatocyte Kit are suitable for a variety of applications in liver research, disease modeling, and hepatotoxicity testing.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    1 KIT
  • Bezeichnung:
    STEMdiff™ Hepatocyte kit
Spezifikationen