ContactUsLinkComponent
MesenCult™ ACF Plus Medium Kit
Catalog # STMC05445
CAS Number:  
undefined
MesenCult™ ACF Plus Medium Kit
Catalog # STMC05445
CAS Number:  
Specifications
  • VE:
    1 KIT
  • Bezeichnung:
    MesenCult™-ACF plus medium kit
Specifications
About this item
Animal component-free medium for human mesenchymal stem cells.
Reduce variability in human mesenchymal stromal cell (MSC; also known as mesenchymal stem cell) cultures and improve experimental reproducibility by using this animal component-free (ACF) and extracellular vesicle (EV)-free medium. MesenCult™-ACF Plus Medium is optimised to derive human MSCs from multiple sources, such as bone marrow or adipose tissue, without serum. Compared to serum-containing or EV-depleted serum-containing media, MSCs cultured with this kit expand more efficiently (rate and cumulative cell total) and without compromising function.
The cultured cells exhibit characteristic MSC surface marker expression and retain robust expansion rate and trilineage differentiation capacities. MesenCult™-ACF Plus Medium Kit is specifically designed for deriving, expanding, and cryopreserving MSCs, as well as differentiating human pluripotent stem cells into mesenchymal progenitors - that is optimised for efficient and consistent MSC cultures. For animal component-free and optimised cryopreservation, MesenCult™-ACF Freezing Medium is recommended for human MSCs previously cultured in MesenCult™ media, including MesenCult™-ACF Plus.
NOTE: Complete MesenCult™-ACF Plus Medium must be supplemented with L-Glutamine. It is also intended to be used in conjunction with Animal Component-Free Cell Attachment Substrate, which is available as part of the MesenCult™-ACF Plus Culture Kit. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').