ContactUsLinkComponent
ImmunoCult™ NK Cell Expansion Coating Material
ImmunoCult™ NK Cell Expansion Coating Material
Catalog # STMC100-0714
CAS Number:  
undefined
ImmunoCult™ NK Cell Expansion Coating Material
Catalog # STMC100-0714
CAS Number:  
Specifications
  • VE:
    1,5 mL
  • Bezeichnung:
    ImmunoCult™ NK cell expansion coating material
Specifications
About this item
Serum-free and animal component-free coating material for expansion of NK cells.
Consistently expand natural killer (NK) cells in serum-free conditions without the use of problematic feeder cells. Use ImmunoCult™ NK Cell Expansion Kit to provide optimised culture conditions for expanding NK cells with high yields. This kit comprises ImmunoCult™ NK Cell Base Medium, ImmunoCult™ NK Cell Expansion Supplement, and ImmunoCult™ NK Cell Expansion Coating Material, providing a complete, easy-to-use culture system. After just 14 days in culture, cells can be harvested and used directly in downstream applications. This kit is compatible with many of our other upstream and downstream products. For example, use EasySep™ cell separation kits to isolate NK cells that can then be immediately expanded using ImmunoCult™ NK Cell Expansion Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').