ContactUsLinkComponent
STEMdiff™ Dorsal Forebrain Organoid Differentiation Kit
Artikelnr. STMC08620
CAS-Nummer:  
undefined
STEMdiff™ Dorsal Forebrain Organoid Differentiation Kit
Artikelnr. STMC08620
CAS-Nummer:  
Spezifikationen
  • VE:
    1 KIT
  • Bezeichnung:
    STEMdiff™ Dorsal forebrain organoid differentiation kit
Spezifikationen
Über diesen Artikel
Cell culture medium kit for robust generation of dorsal forebrain neural organoids from human pluripotent stem cells.
Robustly generate three-dimensional, patterned brain organoid cultures from human pluripotent stem cells without matrix embedding. STEMdiff™ Dorsal and Ventral Forebrain Organoid Differentiation Kits are serum-free cell culture media that work with AggreWell™-generated embryoid bodies to prevent organoid fusion and enable the scalable generation of over 500 highly reproducible organoids per kit. Adapted from protocols, these brain-region-specific organoids are three-dimensional in vitro models with a cellular composition and structural organisation that is representative of the developing human forebrain.
STEMdiff™ Dorsal Forebrain Organoid Differentiation Kit generates tissue of the early developing dorsal pallium, while STEMdiff™ Ventral Forebrain Organoid Differentiation Kit generates tissue of the early developing ventral subpallium. Organoids generated with these kits can also be co-cultured as assembloids to study brain region interactions. For extended periods of organoid culture (>50 days), the components required for organoid maintenance are available as STEMdiff™ Neural Organoid Maintenance Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').