ContactUsLinkComponent
NeuroCult™ Differentiation Kit (Mouse and Rat)
NeuroCult™ Differentiation Kit (Mouse and Rat)
  STMC05704
 :  
undefined
NeuroCult™ Differentiation Kit (Mouse and Rat)
  STMC05704
 :  
  • VE:
    1 KIT
  • Bezeichnung:
    NeuroCult™ Differentiation kit (mouse and rat)
Medium kit for differentiation of mouse and rat neural stem and progenitor cells.
NeuroCult™ Differentiation Kit (Mouse and Rat) is a standardised medium and supplement kit for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').