ContactUsLinkComponent
STEMdiff™ Intestinal Organoid Growth Medium
Catalog # STMC05145
CAS Number:  
undefined
STEMdiff™ Intestinal Organoid Growth Medium
Catalog # STMC05145
CAS Number:  
Specifications
  • VE:
    1 KIT
  • Bezeichnung:
    STEMdiff™ Intestinal organoid growth medium
Specifications
About this item
Serum-free medium kit for maintenance of hPSC-derived human intestinal organoids.
Maintain human intestinal organoids long term as a model system for studying intestinal research, disease modeling, drug discovery, and compound screening. Use serum-free STEMdiff™ Intestinal Organoid Growth Medium (OGM) for extended, consistent culture of human pluripotent stem cell (hPSC)-derived intestinal organoids. Based on the formulation STEMdiff™ Intestinal OGM is optimised for efficient and reproducible organoid generation and expansion across hPSC lines. Use STEMdiff™ Intestinal OGM to mature mid-/hindgut spheroids differentiated from hPSCs using STEMdiff™ Intestinal Organoid Kit. The spheroids will mature into intestinal organoids that you can cryopreserve or maintain long-term through passaging. Intestinal organoids cultured in STEMdiff™ Intestinal OGM exhibit a cellular composition and organisation that models the developing intestinal epithelium and associated mesenchyme, making them a convenient model system with direct relevance to the developing intestine.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').