MegaCult™-C Complete Kit without Cytokines
Artikelnr. STMC04970
Lieferant: STEMCELL Technologies
CAS-Nummer:
Spezifikationen
- VE:1 KIT
- Bezeichnung:MegaCult™ C complete kit without cytokines
Spezifikationen
Über diesen Artikel
Complete kit including collagen, media, staining kit and chamber slide kit for human CFU-Mk assays.
MegaCult™-C Complete Kit without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining. The addition of appropriate cytokines, as chosen by the user, is required.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').