AuftragsverfolgungKontakt
MethoCult™-HY AOF H4034 Medium
MethoCult™-HY AOF H4034 Medium
MethoCult™ H4034 optimum, 100 mL
Catalog # STMC04034
Marketsource
MethoCult™-HY AOF H4034 Medium
Catalog # STMC04034
Marketsource
CAS Number:  
MethoCult™-HY AOF H4034 Medium
MethoCult™-HY AOF H4034 Medium
MethoCult™ H4034 optimum, 100 mL
CAS Number:  
Price
1 420,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Methylcellulose-based medium with recombinant cytokines for human cells. 1×100 ml bottle.
MethoCult™ H4034 Optimum (MethoCult™ GF H4034) is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilised peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4034 Optimum has been formulated to support optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM). The inclusion of G-CSF in Optimum formulations improves discrimination of myeloid colony subtypes. This formulation is compatible with STEMvision™ software for automated colony counting.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • VE:
    100 mL
  • Bezeichnung:
    MethoCult™ H4034 optimum