AuftragsverfolgungKontakt
Alpha MEM with Nucleosides | Avantor
Alpha MEM with Nucleosides
Alpha MEM with nucleosides, 500 mL
Artikelnr. STMC36450
Marketsource
Alpha MEM with Nucleosides
Artikelnr. STMC36450
Marketsource
CAS Nummer:  
Alpha MEM with Nucleosides
Alpha MEM with Nucleosides
Alpha MEM with nucleosides, 500 mL
CAS Nummer:  
Preis
184,00 €
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents
Minimum Essential Medium Eagle - Alpha modification (Alpha MEM) with nucleosides.
Minimum Essential Medium Eagle - Alpha modification (Alpha MEM) with Nucleosides is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition. MEM has typically been used for the cultivation of cells grown in monolayers, however additional supplementation to Alpha MEM has expanded the applications for this medium. MEM was developed by Harry Eagle when Basal Medium Eagle (BME) was found not adequate to support the growth of HeLa cells.
It was discovered that BME could support the growth of a wider variety of cells with the addition of certain medium supplements (e.g. higher concentrations of amino acids), which were incorporated into the MEM formulation. Alpha MEM includes the non-essential amino acids, sodium pyruvate, lipoic acid, ascorbic acid, biotin, and vitamin B12.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    500 mL
  • Bezeichnung:
    Alpha MEM with nucleosides