ContactUsLinkComponent
DermaCult™ Keratinocyte Expansion Medium
DermaCult™ Keratinocyte Expansion Medium
Catalog # STMC100-0500
CAS Number:  
undefined
DermaCult™ Keratinocyte Expansion Medium
Catalog # STMC100-0500
CAS Number:  
Specifications
  • VE:
    1 KIT
  • Bezeichnung:
    DermaCult™ keratinocyte expansion medium
Specifications
About this item
Serum- and BPE-free medium for expansion of primary human keratinocytes.
Improve derivation efficiency and expansion rates for long-term culture of proliferative human primary keratinocytes using BPE- and serum-free DermaCult™ keratinocyte expansion medium and a feeder-free protocol. More efficient cell expansion means that you can perform repeats and technical replicates from the same sample and avoid purchasing additional undifferentiated cells or sourcing patient-derived samples. Unlike other commercial expansion media, DermaCult™ keratinocyte expansion medium enables you to obtain >50 population doublings of human neonatal keratinocytes and >35 population doublings of human adult keratinocytes. With the flexibility to change the media every two days and cryopreserve proliferative cells at any passage - while maintaining their differentiation potential - you can carry out your research at your convenience.
Cells expanded in this medium exhibit cobblestone morphology and express key basal cell markers (K14, p63, ITAG6, and ITGB1) representative of basal keratinocytes and are suitable for differentiation to air-liquid interface (ALI) cultures. Formulated without serum or BPE to maximise experimental reproducibility, confidently use keratinocytes cultured in DermaCult™ for in vitro models to study skin and inflammatory diseases, perform irritation and corrosion assays, or to assess the efficacy of cosmetics or drugs. DermaCult™ keratinocyte expansion medium is compatible for passaging with ACCUTASE™ at 3 to 5 day intervals. It is also suitable for use with collagen, such as to establish keratinocyte culture from skin tissue.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').