AuftragsverfolgungKontakt
STEMdiff™ T Cell Kit | Avantor
STEMdiff™ T Cell Kit
STEMdiff™ T cell kit, 1 KIT
Artikelnr. STMC100-0194
Marketsource
STEMdiff™ T Cell Kit
Artikelnr. STMC100-0194
Marketsource
CAS Nummer:  
STEMdiff™ T Cell Kit
STEMdiff™ T Cell Kit
STEMdiff™ T cell kit, 1 KIT
CAS Nummer:  
Preis
3 150,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
For expansion and differentiation of hPSCs to T cells.
Differentiate human pluripotent stem cells (hPSCs) to T cells with the feeder-free and serum-free STEMdiff™ T cell kit. The STEMdiff™ T Cell Kit protocol first uses animal component-free STEMdiff™ Hematopoietic - EB reagents for the generation of embryoid bodies (EBs) from hPSCs which are then further differentiated into CD34+ cells.
The kit includes all the EB reagents necessary for this step:
STEMdiff™ hematopoietic - EB basal medium
STEMdiff™ hematopoietic - EB supplement A
STEMdiff™ hematopoietic - EB supplement B
The CD34+ cells are then further directed to differentiate to T cells using StemSpan™ reagents:
StemSpan™ SFEM II
StemSpan™ lymphoid progenitor expansion supplement (10X)
StemSpan™ lymphoid differentiation coating material (100X)
StemSpan™ T cell progenitor maturation supplement (10X)
For convenience, all STEMdiff™ Hematopoietic - EB and StemSpan™ reagents included with the kit are also available individually.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    1 KIT
  • Bezeichnung:
    STEMdiff™ T cell kit
Spezifikationen
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.