STEMdiff™ Hematopoietic - EB Basal Medium
STMC100-0171
:
- VE:120 mL
- Bezeichnung:STEMdiff™ hematopoietic-EB basal medium
Medium for the generation of embryoid bodies for downstream lymphoid differentiation.
Use STEMdiff™ hematopoietic - EB basal medium in combination with STEMdiff™ Hematopoietic - EB supplement A and STEMdiff™ hematopoietic - EB supplement B to support the differention of human pluripotent stem cells (hPSCs) to embroyid bodies. Available either individually or as part of the STEMdiff™ NK cell kit or STEMdiff™ T cell kit, where it can be used to promote the differentiation of hPSCs to NK or T cells, respectively.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').