AuftragsverfolgungKontakt
StemSpan™-XF Medium
StemSpan™-XF, 500 mL
Artikelnr. STMC100-0073
Marketsource
StemSpan™-XF Medium
Artikelnr. STMC100-0073
Marketsource
CAS Nummer:  
StemSpan™-XF Medium
StemSpan™-XF Medium
StemSpan™-XF, 500 mL
CAS Nummer:  
Preis
1 510,00 €
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents
Xeno-free and serum-free medium for culture and expansion of human hematopoietic cells.
StemSpan™-XF has been developed for the in vitro culture and expansion of human hematopoietic cells, when the appropriate growth factors and supplements are added. This allows users the flexibility to prepare medium that meets their requirements. StemSpan™-XF contains pre-tested human-derived and recombinant human (rh) proteins. Using appropriate StemSpan™ expansion supplements, StemSpan™-XF may be used to expand CD34+ cells isolated from human cord blood, mobilised peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitor cells to generate populations of erythroid, myeloid, or megakaryocyte progenitor cells.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    500 mL
  • Bezeichnung:
    StemSpan™-XF
Spezifikationen