ContactUsLinkComponent
StemSpan™ Lymphoid Progenitor Expansion Supplement (10X)
StemSpan™ Lymphoid Progenitor Expansion Supplement (10X)
Catalog # STMC09915
CAS Number:  
undefined
StemSpan™ Lymphoid Progenitor Expansion Supplement (10X)
Catalog # STMC09915
CAS Number:  
Specifications
  • VE:
    5 mL
  • Bezeichnung:
    StemSpan™ lymphoid progenitor expansion supplement (10X)
Specifications
About this item
Supplement for the expansion and differentiation of lymphoid progenitor cells.
Use StemSpan™ lymphoid progenitor expansion supplement (10X) to selectively promote the expansion and differentiation of CD34+ cells isolated from human cord blood (CB) samples to CD7+CD5+ progenitor T (pro-T) cells. Available either individually or as part of the StemSpan™ T cell generation kit or STEMdiff™ T cell kit, this supplement is intended for use in combination with StemSpan™ SFEM II medium and plates coated with StemSpan™ lymphoid differentiation coating material (100X). For further maturation of pro-T cells to CD4+CD8+ double-positive (DP) T cells, use StemSpan™ T Cell Progenitor Maturation Supplement (10X). For detailed information on protocols for expansion and differentiation of CD34+ cells with StemSpan™.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').