STEMdiff™ Cerebral Organoid Maturation Kit
Catalog # STMC08571
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- VE:1 KIT
- Bezeichnung:STEMdiff™ cerebral organoid maturation kit
Specifications
About this item
Culture medium kit for extended maturation of human cerebral organoids.
Culture pluripotent stem cell (PSC)-derived neural organoids for extended periods (>40 days) with the STEMdiff™ cerebral organoid maturation kit. For your convenience, the maturation kit is fully compatible with the STEMdiff™ cerebral organoid kit, which can be used to generate neural organoids under defined and serum-free conditions. For more information on protocols for organoid culture with STEMdiff™ cerebral organoid maturation kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').