ContactUsLinkComponent
MegaCult™-C C Medium without Cytokines
MegaCult™-C C Medium without Cytokines
Catalog # STMC04900
CAS Number:  
undefined
MegaCult™-C C Medium without Cytokines
Catalog # STMC04900
CAS Number:  
Specifications
  • VE:
    24 Vial
  • Bezeichnung:
    MegaCult™-C medium without Cytokines
Specifications
About this item
Medium without cytokines.
Use MegaCult™-C medium without cytokines for the culture of colony-forming unit-megakaryocyte progenitor cells (CFU-Mk) in human bone marrow, mobilised peripheral blood, and cord blood samples, after addition of appropriate cytokines. Available either individually or as part of the MegaCult™-C collagen and medium without cytokines kit, this medium is suitable for use with CD34+ enriched cells, mononuclear cells, and cells isolated by other purification methods. It is also intended for assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow, after addition of appropriate cytokines. Required MegaCult™-C medium may be used with collagen solution, available either individually or as part of the MegaCult™-C collagen and medium without cytokines kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').