AuftragsverfolgungKontakt
ClonaCell™-HY Liquid HAT Selection Medium | Avantor
ClonaCell™-HY Liquid HAT Selection Medium
ClonaCell™-HY liquid HAT selection medium, 500 mL
Artikelnr. STMC03831
Marketsource
ClonaCell™-HY Liquid HAT Selection Medium
Artikelnr. STMC03831
Marketsource
CAS Nummer:  
ClonaCell™-HY Liquid HAT Selection Medium
ClonaCell™-HY Liquid HAT Selection Medium
ClonaCell™-HY liquid HAT selection medium, 500 mL
CAS Nummer:  
Preis
579,00 €
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents
Hybridoma selection medium, with HAT (serum-containing).
ClonaCell™-HY liquid HAT selection medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells. This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    500 mL
  • Bezeichnung:
    ClonaCell™-HY liquid HAT selection medium
Spezifikationen
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.