ContactUsLinkComponent
StemSpan™ CD34+ Expansion Supplement (10X)
StemSpan™ CD34+ Expansion Supplement (10X)
Catalog # STMC02691
CAS Number:  
undefined
StemSpan™ CD34+ Expansion Supplement (10X)
Catalog # STMC02691
CAS Number:  
Specifications
  • VE:
    10 mL
  • Bezeichnung:
    StemSpan™ CD34+expansion supplement (10X)
Specifications
About this item
Serum-free culture supplement for expansion of human CD34+ hematopoietic cells.
StemSpan™ CD34+ expansion supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion of CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples. StemSpan™ CD34+ Expansion Supplement typically promotes ~40-fold expansion of total nucleated cells in 7-day liquid cultures of CD34+ human cord blood (CB) cells. After one week, ~40% of the cultured cells express CD34, indicative of >10-fold expansion of input CD34+ CB cells. This expansion may be further increased with the addition of small molecules such as UM729. See data tab for more details.
StemSpan™ CD34+ expansion supplement (10X) is intended for use in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').