AuftragsverfolgungKontakt
McCoy's 5A Medium
McCoy's 5A medium, 500 mL
Artikelnr. STMC36350
Marketsource
McCoy's 5A Medium
Artikelnr. STMC36350
Marketsource
CAS Nummer:  
McCoy's 5A Medium
McCoy's 5A Medium
McCoy's 5A medium, 500 mL
CAS Nummer:  
Preis
194,00 €
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents
McCoy's 5A medium.
McCoy's 5A medium is recommended for a wide variety of cell culture applications. Selection of suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition required for the cell culture application.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spezifikationen
Spezifikationen
  • VE:
    500 mL
  • Bezeichnung:
    McCoy's 5A medium
Spezifikationen