Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 560)
Lieferant: AAT BIOQUEST
Zertifikate
Über diesen Artikel
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 560
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Spezifikationen
- Catalog No:
- AATB103800A0
- AATB103800A1
- Antigen-Symbol:
- CD38
- CD38
- Antigen-Name:
- CD38 antigen
- CD38 antigen
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- iFluor® 560
- iFluor® 560
- Klon:
- HIT2
- HIT2
- Reaktivität:
- Human
- Human
- Wirt:
- Mouse
- Mouse
- Isotyp:
- IgG1
- IgG1
- ImmunFluoreszenz:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
- Reinigung:
- Affinity chromatography
- Affinity chromatography
- Kreuzadsorption:
- No
- No
- Lagertemperatur:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Transporttemperatur:
- 20
- 20